Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethyl-1H-pyrazole
3,4-Dimethyl-1H-pyrazole 98%
3,4-Dimethyl-1H-pyrazole 98%
:
Apollo Scientific
3,4-Dimethyl-1H-pyrazole 98%
:
Apollo Scientific
:
2820-37-3
:
:
C₅H₈N₂
:
MFCD00462154
:
96.133 g/mol
:
111 °C/10mmHg
:
55 °C
:
1.109 g/cm³ (25 °C)
:
Ambient
:
429-130-1
:
GHS05
:
GHS07