Order Entry
Export
ContactUsLinkComponent
2,4-Dihydroxy-3-methylbenzaldehyde 98%
Irritant
2,4-Dihydroxy-3-methylbenzaldehyde 98%
Catalog # APOSOR52629-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dihydroxy-3-methylbenzaldehyde 98%
Catalog # APOSOR52629-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :152.1500 g/mol
Melting Pt:148…154 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:6248-20-0
Formula:C₈H₈O₃
MDL Number:MFCD00156883