Order Entry
Export
ContactUsLinkComponent
 
Irritant
Cyclobutane-1,1-dicarboxylic acid ethyl ester
Catalog # APOSOR52464-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Cyclobutane-1,1-dicarboxylic acid ethyl ester
Catalog # APOSOR52464-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :172.18 g/mol
Density:1.117 g/cm³ (25 °C)
Flash Pt:>110 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:54450-84-9
Formula:C₈H₁₂O₄
MDL Number:MFCD10699115