Order Entry
Export
ContactUsLinkComponent
1,1'-Sulphonyldiimidazole 98%
Irritant
1,1'-Sulphonyldiimidazole 98%
Catalog # APOSOR52410-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,1'-Sulphonyldiimidazole 98%
Catalog # APOSOR52410-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :198.20 g/mol
EINECS:230-554-0
 :GHS07
Storage Temperature:Ambient
CAS Number:7189-69-7
Formula:C₆H₆N₄O₂S
MDL Number:MFCD00015893