


3-(3,5-Dichlorophenoxy)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:81028-92-4
:
Formula:C₁₃H₈Cl₂O₂
MDL Number:MFCD00003355
:267.11 g/mol
Boiling Pt:125 °C/0.03mmHg
Melting Pt:48…55 °C
Flash Pt:113 °C
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient