
6-Methyl-2-pyridone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3279-76-3
:
Formula:C₆H₇NO
MDL Number:MFCD00006279
Melting Pt:155…160 °C
Density:1.13 g/cm³ (25 °C)
:109.13 g/mol
EINECS:221-919-5
:GHS07
Storage Temperature:Ambient