11-Bromo-1-undecene 95%
Catalog # APOSOR52369-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:233.19 g/mol
Boiling Pt:149…150 °C
Density:1.063 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
CAS Number:7766-50-9
Formula:C₁₁H₂₁Br
MDL Number:MFCD00040825