Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
9-(4-Bromophenyl)-10-phenylanthracene
9-(4-Bromophenyl)-10-phenylanthracene 98%
9-(4-Bromophenyl)-10-phenylanthracene 98%
APOSOR52348-1G
:
Apollo Scientific
:
9-(4-Bromophenyl)-10-phenylanthracene 98%
APOSOR52348-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
409.33 g/mol
:
227 °C
:
GHS07
:
Ambient
:
625854-02-6
:
C₂₆H₁₇Br
:
MFCD24842536