
10-Undecynoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2777-65-3
:
Formula:C₁₁H₁₈O₂
MDL Number:MFCD00014389
:182.26 g/mol
Boiling Pt:180 °C/15mmHg
Melting Pt:40…42 °C
Flash Pt:113 °C
Storage Temperature:Refrigerator
EINECS:220-471-8
:GHS07