1,3-Diphenylacetone
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
- CAS Number:102-04-5
- Formula:(C₆H₅)CH₂C(O)CH₂(C₆H₅)
- MDL Number:MFCD00004795
- Molecular Weight:210.28 g/mol
- Boiling Pt:330 °C
- Melting Pt:32…34 °C
- Density:1.04 g/cm³ (25 °C)
- Flash Pt:113 °C
- Storage Temperature:Ambient
- EINECS:203-000-0
3 Options Available Below
Product Details & Documents
Synonyms: 1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone
- CAS Number:102-04-5
- Formula:(C₆H₅)CH₂C(O)CH₂(C₆H₅)
- MDL Number:MFCD00004795
- Molecular Weight:210.28 g/mol
- Boiling Pt:330 °C
- Melting Pt:32…34 °C
- Density:1.04 g/cm³ (25 °C)
- Flash Pt:113 °C
- Storage Temperature:Ambient
- EINECS:203-000-0



