
2-Methyl-1,6-naphthyridine-3-carboxylic acid 95+%
Catalog # APOSOR5224-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:188.19 g/mol
Melting Pt:267…269 °C
:GHS07
Storage Temperature:Ambient
CAS Number:387350-63-2
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD00202955