Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloropyridine-2,3-diamine
4-Chloropyridine-2,3-diamine 97%
4-Chloropyridine-2,3-diamine 97%
4-Chloropyridine-2,3-diamine 97%
APOSOR52182-1G
:
Apollo Scientific
:
4-Chloropyridine-2,3-diamine 97%
APOSOR52182-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
143.57 g/mol
:
Ambient
:
24484-98-8
:
C₅H₆ClN₃
:
MFCD13181777