Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol 98%
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol 98%
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
159873-64-0
:
Formula:
C₉H₉NO₅
MDL Number:
MFCD08274663
:
211.17 g/mol
Storage Temperature:
Ambient