Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Dichloro-9,10-anthraquinone
1,5-Dichloro-9,10-anthraquinone 98%
1,5-Dichloro-9,10-anthraquinone 98%
APOSOR52154-100G
:
Apollo Scientific
:
1,5-Dichloro-9,10-anthraquinone 98%
APOSOR52154-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
277.11 g/mol
:
245…247 °C
:
201-424-0
:
GHS07
:
Ambient
:
82-46-2
:
C₁₄H₆Cl₂O₂
:
MFCD00001190