Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloroquinazoline
2,4-Dichloroquinazoline 97%
2,4-Dichloroquinazoline 97%
Irritant
2,4-Dichloroquinazoline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Dichloroquinazoline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
607-68-1
:
Formula:
C₈H₄Cl₂N₂
MDL Number:
MFCD00091950
:
199.04 g/mol
Melting Pt:
117…119 °C
:
GHS07
Storage Temperature:
Ambient