Order Entry
Export
ContactUsLinkComponent
Methyl-p-toluate 97%
Irritant
Methyl-p-toluate 97%
Catalog # APOSOR52042-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-p-toluate 97%
Catalog # APOSOR52042-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :150.18
Boiling Pt:103…104 °C/15mmHg
Melting Pt:32…36 °C
Density:1.06 g/cm³ (25 °C)
Flash Pt:90 °C
Storage Temperature:Ambient
EINECS:202-784-1
 :GHS07
CAS Number:99-75-2
Formula:C₉H₁₀O₂
MDL Number:MFCD00008441