0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,4-Dichloro-6-phenyl-1,3,5-triazine
2,4-Dichloro-6-phenyl-1,3,5-triazine 98%
2,4-Dichloro-6-phenyl-1,3,5-triazine 98%
:
Apollo Scientific
2,4-Dichloro-6-phenyl-1,3,5-triazine 98%
:
Apollo Scientific
:
1700-02-3
:
:
C₉H₅Cl₂N₃
:
MFCD00047340
:
226.0600 g/mol
:
GHS07
:
Refrigerator