0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4,6-Dichloropyrimidine-5-carbonitrile
4,6-Dichloropyrimidine-5-carbonitrile 97%
4,6-Dichloropyrimidine-5-carbonitrile 97%
Irritant
4,6-Dichloropyrimidine-5-carbonitrile 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,6-Dichloropyrimidine-5-carbonitrile 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
5305-45-3
:
Formula:
C₅HCl₂N₃
MDL Number:
MFCD08460393
:
173.9875 g/mol
:
GHS07
Storage Temperature:
Refrigerator