Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dithiane 98%

1,3-Dithiane 98%

Supplier:   Apollo Scientific
1,3-Dithiane 98%
Supplier:  Apollo Scientific
Certificates
1,3-Dithiane 98%

6 Options Available Below

Product Details & Documents
  • CAS Number:505-23-7
  • Formula:C₄H₈S₂
  • MDL Number:MFCD00006654
  • Molecular Weight:120.24 g/mol
  • Boiling Pt:207…208 °C
  • Melting Pt:52…54 °C
  • Flash Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:208-006-7
  • UN:3335
  • ADR:9,III
  •  :Density: 1.36 g/cm³ (25 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 47,80 €
    47,80 €
    1 * 25 g
       47,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 184,00 €
    184,00 €
    1 * 100 g
       184,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 915,00 €
    915,00 €
    1 * 500 g
       915,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 4 570,00 €
    4 570,00 €
    1 * 2,5 kg
       4 570,00 €

    Specifications:
    New Product
    New Product