


Bis[3-(trimethoxysilyl)propyl]amine
Catalog # APOSOR51899-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:341.55 g/mol
Boiling Pt:152 °C/3.75mmHg
Density:1.04 g/cm³ (25 °C)
Flash Pt:110 °C
Storage Temperature:Ambient
UN:3082
ADR:9+EHS,III
:GHS05
:GHS07
:GHS09
CAS Number:82985-35-1
Formula:C₁₂H₃₁NO₆Si₂
MDL Number:MFCD00191835