
2,6-Dimethoxypyrazine
Catalog # APOSOR51827-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:75 °C/10mmHg
Melting Pt:48…49 °C
:140.14 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:4774-15-6
Formula:C₆H₈N₂O₂
MDL Number:MFCD00955843