
2,3-Dichlorobenzoyl chloride
Catalog # APOSOR5179-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:209.46 g/mol
Boiling Pt:140 °C/14mmHg
Melting Pt:28…32 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:220-811-5
UN:3261
ADR:8,II
:GHS05
CAS Number:2905-60-4
Formula:C₇H₃Cl₃O
MDL Number:MFCD00035937