


N,O-Bis(trimethylsilyl)acetamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10416-59-8
:
Formula:C₈H₂₁NOSi₂
MDL Number:MFCD00008270
:203.43 g/mol
Boiling Pt:71…73 °C/35mmHg
Melting Pt:24 °C
Density:0.832 g/cm³ (25 °C)
Flash Pt:42 °C
Storage Temperature:Refrigerator
EINECS:233-892-7
UN:2920
ADR:8+3,II
:GHS02
:GHS05
:GHS07