1,4-Dimethylpiperazine
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: sym-Dimethylpiperazine
- CAS Number:106-58-1
- Formula:C₆H₁₄N₂
- MDL Number:MFCD00006156
- Molecular Weight:114.19 g/mol
- Boiling Pt:131…132 °C/750mmHg
- Melting Pt:–1 °C
- Density:0.844 g/cm³ (25 °C)
- Flash Pt:18 °C
- Storage Temperature:Ambient
- EINECS:203-412-0
- UN:2924
- ADR:3+8,II
3 Options Available Below
Product Details & Documents
Synonyms: sym-Dimethylpiperazine
- CAS Number:106-58-1
- Formula:C₆H₁₄N₂
- MDL Number:MFCD00006156
- Molecular Weight:114.19 g/mol
- Boiling Pt:131…132 °C/750mmHg
- Melting Pt:–1 °C
- Density:0.844 g/cm³ (25 °C)
- Flash Pt:18 °C
- Storage Temperature:Ambient
- EINECS:203-412-0
- UN:2924
- ADR:3+8,II



