
2-Chloro-3,5-dinitropyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2578-45-2
:
Formula:C₅H₂ClN₃O₄
MDL Number:MFCD00006233
:203.54 g/mol
Melting Pt:63…66 °C
Storage Temperature:Ambient
EINECS:219-937-3
UN:2811
ADR:6.1,II
:GHS06