Order Entry
Export
ContactUsLinkComponent
6,6'-Dicyano-2,2'-bipyridyl 97%
6,6'-Dicyano-2,2'-bipyridyl 97%
Catalog # APOSOR50969-250MG
Supplier:  Apollo Scientific
CAS Number:  
6,6'-Dicyano-2,2'-bipyridyl 97%
Catalog # APOSOR50969-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :206.21 g/mol
Storage Temperature:Ambient
CAS Number:4411-83-0
Formula:C₁₂H₆N₄
MDL Number:MFCD00797251