Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dibromo-2,1,3-benzothiadiazole
5,6-Dibromo-2,1,3-benzothiadiazole
5,6-Dibromo-2,1,3-benzothiadiazole
Irritant
5,6-Dibromo-2,1,3-benzothiadiazole
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5,6-Dibromo-2,1,3-benzothiadiazole
Supplier:
Apollo Scientific
About this item
CAS Number:
18392-81-9
:
Formula:
C₆H₂Br₂N₂S
MDL Number:
MFCD20257910
:
293.9700 g/mol
Melting Pt:
132…133 °C
:
GHS07
Storage Temperature:
Ambient