Order Entry
Export
ContactUsLinkComponent
7-Hydroxycitronellal 95%
7-Hydroxycitronellal 95%
Catalog # APOSOR50940-100G
Supplier:  Apollo Scientific
CAS Number:  
7-Hydroxycitronellal 95%
Catalog # APOSOR50940-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :172.2646 g/mol
Boiling Pt:257 °C
Density:0.92 g/cm³ (25 °C)
Storage Temperature:Freezer
CAS Number:107-75-5
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00014681