
2,3-Dimethoxyphenol 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5150-42-5
:
Formula:C₈H₁₀O₃
MDL Number:MFCD00008366
:154.17 g/mol
Boiling Pt:233…234 °C
Density:1.182 g/cm³ (25 °C)
Flash Pt:109 °C
EINECS:225-922-2
:GHS07
Storage Temperature:Ambient