3,3-Diphenylpropylamine 98%
Supplier: Apollo Scientific
About this item
Synonyms: 3,3-Diphenylpropan-1-amine
- CAS Number:5586-73-2
- Formula:C₁₅H₁₇N
- MDL Number:MFCD00008202
- Molecular Weight:211.31 g/mol
- Boiling Pt:166 °C/2mmHg
- Melting Pt:24…31 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:226-984-3
3 Options Available Below
Product Details & Documents
Synonyms: 3,3-Diphenylpropan-1-amine
- CAS Number:5586-73-2
- Formula:C₁₅H₁₇N
- MDL Number:MFCD00008202
- Molecular Weight:211.31 g/mol
- Boiling Pt:166 °C/2mmHg
- Melting Pt:24…31 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:226-984-3
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.APOSOR5085-5G - Specifications:
More
Specifications
Cat. No.APOSOR5085-25G - Specifications:
More
Specifications
Cat. No.APOSOR5085-100G



