Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Cyclohexanedione
1,4-Cyclohexanedione 98%
1,4-Cyclohexanedione 98%
:
Apollo Scientific
1,4-Cyclohexanedione 98%
:
Apollo Scientific
:
637-88-7
:
:
C₆H₈O₂
:
MFCD00001606
:
112 °C/19.5mmHg
:
76…80 °C
:
132 °C
:
112.13 g/mol
:
Refrigerator
:
211-306-0
:
GHS07