

4-Chloromethyl-o-xylene 70%
Catalog # APOSOR5034-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
Specifications
About this item
:
Boiling Pt:117 °C/24mmHg
Density:1.06 g/cm³ (25 °C)
EINECS:203-032-5
UN:3265
ADR:8,II
CAS Number:102-46-5