Order Entry
Export
Orders LinkContactUsLinkComponent
α-Chloro-3-nitrotoluene 96%

α-Chloro-3-nitrotoluene 96%

Bottle, 10 g
Catalog # APOSOR5032-10G
Corrosive
Supplier:   Apollo Scientific
α-Chloro-3-nitrotoluene 96%
Catalog # APOSOR5032-10G
Supplier:  Apollo Scientific
CAS Number:  619-23-8
α-Chloro-3-nitrotoluene 96%

Bottle, 10 g

CAS Number:  619-23-8
Price
21,80 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  1-(Chloromethyl)-3-nitrobenzene, 3-Nitrobenzyl chloride, α-Chloro-m-nitrotoluene, alpha-Chloro-3-nitrotoluene, m-Nitrobenzyl chloride
  • CAS Number:619-23-8
  • Formula:C₇H₆ClNO₂
  • MDL Number:MFCD00007272
  • Molecular Weight:171.58 g/mol
  • Boiling Pt:85…87 °C/5mmHg
  • Melting Pt:43…47 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:210-586-1
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications