
2,4-Dichloro-6-methoxyaniline 95+%
Catalog # APOSOR500006-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:192.04 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:93839-14-6
Formula:C₇H₇Cl₂NO
MDL Number:MFCD20693704