0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
5-(3,4-Dichlorophenyl)isoxazole
5-(3,4-Dichlorophenyl)isoxazole
5-(3,4-Dichlorophenyl)isoxazole
APOSOR5-250MG
:
Apollo Scientific
:
5-(3,4-Dichlorophenyl)isoxazole
APOSOR5-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
GHS07
:
Ambient
:
138716-63-9
:
C₉H₅Cl₂NO