3,5-Dimethoxybenzaldehyde 97%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:7311-34-4
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00003366
- Molecular Weight:166.18 g/mol
- Boiling Pt:115…118 °C/1mmHg
- Melting Pt:44…48 °C
- Storage Temperature:Ambient
6 Options Available Below
Product Details & Documents
- CAS Number:7311-34-4
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00003366
- Molecular Weight:166.18 g/mol
- Boiling Pt:115…118 °C/1mmHg
- Melting Pt:44…48 °C
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



