2-Nitrobenzaldehyde 98%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:552-89-6
- Formula:C₇H₅NO₃
- MDL Number:MFCD00007132
- Molecular Weight:151.12 g/mol
- Boiling Pt:153 °C/23mmHg
- Melting Pt:42…46 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:209-025-3
2 Options Available Below
Product Details & Documents
- CAS Number:552-89-6
- Formula:C₇H₅NO₃
- MDL Number:MFCD00007132
- Molecular Weight:151.12 g/mol
- Boiling Pt:153 °C/23mmHg
- Melting Pt:42…46 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:209-025-3



