1,4-Benzodioxane
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan
- CAS Number:493-09-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00006821
- Molecular Weight:136.15 g/mol
- Boiling Pt:216 °C
- Density:1.17 g/cm³ (25 °C)
- Flash Pt:87 °C
- Storage Temperature:Ambient
- EINECS:207-775-6
6 Options Available Below
Product Details & Documents
Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan
- CAS Number:493-09-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00006821
- Molecular Weight:136.15 g/mol
- Boiling Pt:216 °C
- Density:1.17 g/cm³ (25 °C)
- Flash Pt:87 °C
- Storage Temperature:Ambient
- EINECS:207-775-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



