1,3-Benzodioxole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:274-09-9
:
Formula:C₇H₆O₂
MDL Number:MFCD00005818
:122.12 g/mol
Boiling Pt:172…173 °C
Melting Pt:–18 °C
Density:1.185 g/cm³ (25 °C)
Flash Pt:55 °C
Storage Temperature:Ambient
EINECS:205-992-0
UN:1993
ADR:3,III
:GHS02
:GHS07