Order Entry
Export
ContactUsLinkComponent
4,4'-Methylenedi-o-toluidine
4,4'-Methylenedi-o-toluidine
Catalog # APOSOR49083-25G
Supplier:  Apollo Scientific
CAS Number:  
4,4'-Methylenedi-o-toluidine
Catalog # APOSOR49083-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :226.3200 g/mol
Melting Pt:156 °C
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
CAS Number:838-88-0
Formula:C₁₅H₁₈N₂
MDL Number:MFCD00126963