1,2-Diaminocyclohexane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:694-83-7
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00001491
:114.19 g/mol
Boiling Pt:188…192 °C
Melting Pt:42036 °C
Flash Pt:75 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III