Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Piperidine-2,4-dione
Piperidine-2,4-dione 97%
Piperidine-2,4-dione 97%
Irritant
Piperidine-2,4-dione 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Piperidine-2,4-dione 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
50607-30-2
:
Formula:
C₅H₇NO₂
MDL Number:
MFCD08704814
:
113.1146 g/mol
Melting Pt:
98…103 °C
Storage Temperature:
Ambient
UN:
3335
ADR:
9,III
:
GHS07