Order Entry
Export
ContactUsLinkComponent
3,5-Dibenzyloxybenzoic acid 96%
3,5-Dibenzyloxybenzoic acid 96%
Catalog # APOSOR4816-25G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dibenzyloxybenzoic acid 96%
Catalog # APOSOR4816-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :334.37 g/mol
Storage Temperature:Ambient
CAS Number:28917-43-3
Formula:C₂₁H₁₈O₄
MDL Number:MFCD00191341