Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromo-2,6-dichlorophenol 95%

4-Bromo-2,6-dichlorophenol 95%

Bottle
Catalog # APOSOR480798-5G
New Product
Supplier:   Apollo Scientific
4-Bromo-2,6-dichlorophenol 95%
Catalog # APOSOR480798-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  3217-15-0
4-Bromo-2,6-dichlorophenol 95%

Bottle

CAS Number:  3217-15-0
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:3217-15-0
  • Formula:C₆H₃BrCl₂O
  • MDL Number:MFCD00019996
  • Molecular Weight:241.9 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications