Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloropyrimidine-5-carbonitrile
2,4-Dichloropyrimidine-5-carbonitrile 97%
2,4-Dichloropyrimidine-5-carbonitrile 97%
APOSOR480750-5G
:
Apollo Scientific
:
2,4-Dichloropyrimidine-5-carbonitrile 97%
APOSOR480750-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
173.9875 g/mol
:
Ambient
:
3177-24-0
:
C₅HCl₂N₃
:
MFCD09746248