Order Entry
Export
ContactUsLinkComponent
2,6-Dihydroxynaphthalene 97%
2,6-Dihydroxynaphthalene 97%
Catalog # APOSOR480646-25G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dihydroxynaphthalene 97%
Catalog # APOSOR480646-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :160.17 g/mol
Melting Pt:220 °C
Storage Temperature:Ambient
CAS Number:581-43-1
Formula:C₁₀H₈O₂
MDL Number:MFCD00004082