Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-1,3-dimethyluracil
6-Chloro-1,3-dimethyluracil 98%
6-Chloro-1,3-dimethyluracil 98%
APOSOR480621-25G
:
Apollo Scientific
:
6-Chloro-1,3-dimethyluracil 98%
APOSOR480621-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
174.59 g/mol
:
109…113 °C
:
Ambient
:
6972-27-6
:
C₆H₇ClN₂O₂
:
MFCD00038066