Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromopyrazolo[1,5-a]pyrimidine
3-Bromopyrazolo[1,5-a]pyrimidine 97%
3-Bromopyrazolo[1,5-a]pyrimidine 97%
3-Bromopyrazolo[1,5-a]pyrimidine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromopyrazolo[1,5-a]pyrimidine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
55405-67-9
:
Formula:
C6H4BrN3
MDL Number:
MFCD09832893
:
198.0201 g/mol
Storage Temperature:
Ambient