Order Entry
Export
ContactUsLinkComponent
Methyl-2-methylnicotinate 97%
Methyl-2-methylnicotinate 97%
Catalog # APOSOR480568-25G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2-methylnicotinate 97%
Catalog # APOSOR480568-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :151.16 g/mol
Storage Temperature:Ambient
CAS Number:65719-09-7
Formula:C₈H₉NO₂
MDL Number:MFCD00051961