2,4-Dihydroxy-5-methoxypyrimidine 95%
Catalog # APOSOR480522-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:142.1100 g/mol
Storage Temperature:Ambient
CAS Number:6623-81-0
Formula:C₅H₆N₂O₃
MDL Number:MFCD00187903